Assay Details
Gene Symbol: NNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006169.2 | NP_006160.1 | 3 | 1484 | 99 | |
GenBank mRNA | AK026845.1 | - | 3 | 861 | 99 | |
BC000234.1 | - | 5 | 1004 | 3350358 | 99 | |
CR541907.1 | - | 3 | 740 | 99 | ||
U08021.1 | - | 3 | 857 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000627588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000627588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
