Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000312960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000472885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
ENST00000482706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012807.hg.1 |
NM_005067.5 |
Refseq |