Assay Details
Gene Symbol: TRIM16L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005256481.4 | XP_005256538.1 | 5 | 94 | ||
XM_005256483.4 | XP_005256540.1 | 4 | 94 | |||
XM_011523690.2 | XP_011521992.1 | 5 | 94 | |||
XM_017024245.1 | XP_016879734.1 | 5 | 94 | |||
XM_017024246.1 | XP_016879735.1 | 5 | 94 | |||
XM_017024247.1 | XP_016879736.1 | 5 | 565 | 97 | ||
XM_017024248.1 | XP_016879737.1 | 4 | 94 | |||
XM_017024249.1 | XP_016879738.1 | 4 | 94 | |||
XM_017024250.1 | XP_016879739.1 | 4 | 455 | 97 | ||
XM_017024251.1 | XP_016879740.1 | 4 | 94 | |||
GenBank mRNA | AL080124.1 | - | 4 | 94 | ||
BC127827.1 | - | 4 | 40131713 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC127827.1, XM_005256481.4, XM_005256483.4, XM_011523690.2, XM_017024245.1, XM_017024246.1, XM_017024247.1, XM_017024248.1, XM_017024249.1, XM_017024250.1, XM_017024251.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
