Assay Details
Gene Symbol: BDKRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000623.3 | NP_000614.1 | 3 | 80 | ||
GenBank mRNA | AI095360.1 | - | 1 | 200 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
