Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000475512.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000475512.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000216068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000406199.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000475512.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000475512.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
ENST00000486019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009355.hg.1 |
NM_005740.2 |
Refseq |