Assay Details
Gene Symbol: MYO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005254397.3 | XP_005254454.1 | 32 | 4296 | 107 | |
XM_011521606.2 | XP_011519908.2 | 32 | 4299 | 107 | ||
XM_011521607.2 | XP_011519909.2 | 31 | 4290 | 107 | ||
XM_011521609.2 | XP_011519911.2 | 31 | 4217 | 107 | ||
XM_017022227.1 | XP_016877716.1 | 31 | 4191 | 107 | ||
GenBank mRNA | AF055459.1 | - | 1 | 150 | 107 | |
AF090424.1 | - | 5 | 599 | 107 | ||
AF090425.1 | - | 4 | 590 | 107 | ||
AF090427.1 | - | 4 | 518 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
