Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000329610.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
ENST00000566435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011328.hg.1 |
NM_001099456.2 |
Refseq |