Assay Details
Gene Symbol: MYH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256012.1 | NP_001242941.1 | 24 | 3016 | 98 | |
NM_001256095.1 | NP_001243024.1 | 23 | 2950 | 98 | ||
NM_005964.3 | NP_005955.3 | 22 | 2923 | 98 | ||
XM_011523875.2 | XP_011522177.1 | 24 | 3305 | 98 | ||
XM_011523878.2 | XP_011522180.1 | 23 | 3294 | 98 | ||
XM_011523879.2 | XP_011522181.1 | 23 | 3239 | 98 | ||
XM_011523880.2 | XP_011522182.1 | 22 | 3212 | 98 | ||
XM_017024677.1 | XP_016880166.1 | 23 | 3008 | 98 | ||
XM_017024678.1 | XP_016880167.1 | 24 | 2990 | 98 | ||
XM_017024679.1 | XP_016880168.1 | 24 | 3031 | 98 | ||
XM_017024680.1 | XP_016880169.1 | 24 | 3339 | 98 | ||
XM_017024681.1 | XP_016880170.1 | 23 | 2960 | 98 | ||
XM_017024682.1 | XP_016880171.1 | 23 | 2935 | 98 | ||
GenBank mRNA | AB210026.1 | - | 24 | 2968 | 98 | |
BC008968.2 | - | 6 | 627 | 98 | ||
BC117690.1 | - | 21 | 2825 | 40072059 | 98 | |
BC117691.1 | - | 21 | 2825 | 40072061 | 98 | |
BC144668.1 | - | 23 | 2907 | 9053199 | 98 | |
BC150634.1 | - | 23 | 2907 | 9057004 | 98 | |
M69181.1 | - | 22 | 2863 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
