Assay Details
Gene Symbol: PHYH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037537.1 | NP_001032626.1 | 7 | 1113 | 74 | |
NM_001323080.1 | NP_001310009.1 | 8 | 1172 | 74 | ||
NM_001323082.1 | NP_001310011.1 | 8 | 1014 | 74 | ||
NM_001323083.1 | NP_001310012.1 | 6 | 744 | 74 | ||
NM_001323084.1 | NP_001310013.1 | 7 | 1119 | 74 | ||
NM_006214.3 | NP_006205.1 | 8 | 1008 | 74 | ||
GenBank mRNA | AF023462.1 | - | 8 | 947 | 74 | |
AF112977.1 | - | 8 | 979 | 74 | ||
BC029512.1 | - | 8 | 968 | 5277185 | 74 | |
CR542055.1 | - | 8 | 920 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
