Assay Details
Gene Symbol: TISP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY713301.1 | - | 2 | 271 | 86 |
Gene Symbol: LOC646743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY726562.1 | - | 2 | 296 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
