Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000296233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
ENST00000509675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012275.hg.1 |
NM_014079.3 |
Refseq |