Assay Details
Gene Symbol: NPM1-ALK fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_NPM1-ALK_N4A20_COSF198.0 | - | 59 | 89 |
Gene Symbol: NPM1-ALK fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D45915.1 | - | 417 | 89 | ||
U04946.1 | - | 328 | 89 |
