Assay Details
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006719749.3 | XP_006719812.1 | 63 | 13989 | 103 | |
XM_017020303.1 | XP_016875792.1 | 63 | 14466 | 103 | ||
XM_017020304.1 | XP_016875793.1 | 62 | 12890 | 103 | ||
XM_017020305.1 | XP_016875794.1 | 63 | 14485 | 103 | ||
XM_017020306.1 | XP_016875795.1 | 42 | 9627 | 103 | ||
XM_017020307.1 | XP_016875796.1 | 36 | 8980 | 103 | ||
GenBank mRNA | BC036296.1 | - | 1 | 547 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
