Assay Details
Gene Symbol: SLC24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254740.1 | NP_001241669.1 | 7 | 3124 | 132 | |
NM_001301031.1 | NP_001287960.1 | 8 | 5111 | 132 | ||
NM_001301032.1 | NP_001287961.1 | 8 | 5147 | 132 | ||
NM_004727.2 | NP_004718.1 | 10 | 5362 | 132 | ||
XM_011522220.2 | XP_011520522.1 | 10 | 5282 | 132 | ||
XM_011522221.2 | XP_011520523.1 | 10 | 5246 | 132 | ||
GenBank mRNA | AI026072.1 | - | 1 | 239 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
