Assay Details
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019020.3 | NP_061893.2 | 4 | 946 | 147 | |
XM_005257049.2 | XP_005257106.1 | 4 | 957 | 147 | ||
XM_005257050.3 | XP_005257107.1 | 4 | 963 | 147 | ||
XM_005257052.3 | XP_005257109.1 | 4 | 963 | 147 | ||
XM_006721694.3 | XP_006721757.1 | 4 | 963 | 147 | ||
XM_006721695.3 | XP_006721758.1 | 4 | 963 | 147 | ||
XM_011524322.2 | XP_011522624.1 | 4 | 963 | 147 | ||
XM_011524323.2 | XP_011522625.2 | 2 | 855 | 147 | ||
XM_017024185.1 | XP_016879674.1 | 4 | 963 | 147 | ||
XM_017024186.1 | XP_016879675.1 | 2 | 698 | 147 | ||
XM_017024187.1 | XP_016879676.1 | 3 | 922 | 147 | ||
XM_017024188.1 | XP_016879677.1 | 2 | 78 | 147 | ||
GenBank mRNA | AB449902.1 | - | 3 | 820 | 147 | |
BC028290.1 | - | 4 | 936 | 4480654 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017024188.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
