Assay Details
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011530302.2 | XP_011528604.1 | 1 | 1788 | 266 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530302.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
