Assay Details
Gene Symbol: TPCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137479.1 | - | 15 | 3035 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
