Assay Details
Gene Symbol: HIST1H2APS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045125.1 | 1 | 226 | 97 | ||
GenBank mRNA | DB441866.1 | - | 1 | 226 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
