Assay Details
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK092374.1 | - | 1 | 2993 | 115 | |
AK093865.1 | - | 1 | 1252 | 115 | ||
BX537651.1 | - | 1 | 1362 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
