Assay Details
Gene Symbol: FTX | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028379.1 | 4 | 564 | 73 | |||
GenBank mRNA | AK057701.1 | - | LIT1007 Ftx |
4 | 515 | 73 | |
AK311345.1 | - | 4 | 564 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
