Assay Details
Gene Symbol: ALG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724693.2 | XP_006724756.1 | 1 | 284 | 69 | |
XM_006724697.3 | XP_006724760.2 | 1 | 288 | 69 | ||
XM_006724698.3 | XP_006724761.2 | 1 | 288 | 69 | ||
XM_011531028.2 | XP_011529330.1 | 1 | 285 | 69 | ||
XM_011531030.1 | XP_011529332.1 | 1 | 283 | 69 | ||
XM_017029846.1 | XP_016885335.1 | 1 | 286 | 69 | ||
XM_017029847.1 | XP_016885336.1 | 1 | 285 | 69 | ||
XM_017029848.1 | XP_016885337.1 | 1 | 283 | 69 | ||
XM_017029849.1 | XP_016885338.1 | 1 | 284 | 69 | ||
XM_017029850.1 | XP_016885339.1 | 1 | 283 | 69 | ||
XM_017029851.1 | XP_016885340.1 | 1 | 286 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006724693.2, XM_006724697.3, XM_006724698.3, XM_011531028.2, XM_011531030.1, XM_017029846.1, XM_017029847.1, XM_017029848.1, XM_017029849.1, XM_017029850.1, XM_017029851.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
