Assay Details
Gene Symbol: ALG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724693.2 | XP_006724756.1 | 27 | 4622 | 111 | |
XM_006724697.3 | XP_006724760.2 | 28 | 4633 | 111 | ||
XM_006724698.3 | XP_006724761.2 | 28 | 4758 | 111 | ||
XM_011531028.2 | XP_011529330.1 | 28 | 4734 | 111 | ||
XM_011531030.1 | XP_011529332.1 | 27 | 4495 | 111 | ||
XM_011531031.1 | XP_011529333.1 | 28 | 4433 | 111 | ||
XM_011531032.1 | XP_011529334.1 | 28 | 4239 | 111 | ||
XM_011531033.2 | XP_011529335.1 | 28 | 4279 | 111 | ||
XM_011531034.2 | XP_011529336.1 | 26 | 3973 | 111 | ||
XM_011531037.1 | XP_011529339.1 | 18 | 2990 | 111 | ||
XM_017029846.1 | XP_016885335.1 | 28 | 4711 | 111 | ||
XM_017029847.1 | XP_016885336.1 | 28 | 4695 | 111 | ||
XM_017029848.1 | XP_016885337.1 | 26 | 4399 | 111 | ||
XM_017029851.1 | XP_016885340.1 | 29 | 4641 | 111 | ||
XM_017029852.1 | XP_016885341.1 | 21 | 3306 | 111 | ||
XM_017029853.1 | XP_016885342.1 | 24 | 3548 | 111 | ||
XM_017029854.1 | XP_016885343.1 | 15 | 3070 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
