Assay Details
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK058006.1 | - | 1 | 1661 | 194 | |
AK093908.1 | - | 1 | 980 | 194 | ||
AY423733.1 | - | 1 | 1673 | 194 | ||
AY871271.1 | - | 1 | 995 | 194 | ||
BX537651.1 | - | 1 | 3206 | 194 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
