Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152243.2 | NP_689449.1 | 3 | 2029 | 117 | |
XM_006724107.1 | XP_006724170.1 | 3 | 2027 | 117 | ||
XM_006724108.1 | XP_006724171.1 | 3 | 2033 | 117 | ||
XM_006724109.1 | XP_006724172.1 | 3 | 1993 | 117 | ||
XM_017028541.1 | XP_016884030.1 | 3 | 2044 | 117 | ||
GenBank mRNA | BC009356.2 | - | 4 | 1978 | 4300677 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
