Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085400.1 | NP_001078869.1 | 7 | 2494 | 97 | |
XM_017008590.1 | XP_016864079.1 | 5 | 2162 | 97 | ||
GenBank mRNA | AK025431.1 | - | 3 | 931 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
