Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005252.3 | NP_005243.1 | 1 | 215 | 59 | |
GenBank mRNA | AK097379.1 | - | 1 | 165 | 59 | |
AK291326.1 | - | 1 | 165 | 59 | ||
AK298659.1 | - | 1 | 165 | 59 | ||
BC004490.2 | - | 1 | 136 | 3688670 | 59 | |
BX647104.1 | - | 1 | 165 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK097379.1, AK291326.1, AK298659.1, BC004490.2, NM_005252.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
