Assay Details
Gene Symbol: LOC283278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL833334.1 | - | 1 | 2767 | 68 | |
AL833346.1 | - | 1 | 2767 | 68 |
Gene Symbol: PLEKHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC032761.1 | - | 1 | 473 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
