Assay Details
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090452.1 | - | 1 | 395 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
