Assay Details
Gene Symbol: NDRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022477.3 | NP_071922.2 | 15 | 113 | ||
NM_032013.3 | NP_114402.1 | 16 | 113 | |||
NR_038370.1 | 14 | 113 | ||||
XM_006723837.3 | XP_006723900.1 | 17 | 113 | |||
XM_006723838.3 | XP_006723901.1 | 15 | 113 | |||
XM_011528925.2 | XP_011527227.1 | 15 | 113 | |||
XM_011528926.2 | XP_011527228.1 | 14 | 113 | |||
XM_011528927.2 | XP_011527229.1 | 16 | 113 | |||
XM_011528928.2 | XP_011527230.1 | 15 | 113 | |||
XM_017027978.1 | XP_016883467.1 | 17 | 113 | |||
XM_017027979.1 | XP_016883468.1 | 16 | 113 | |||
XM_017027980.1 | XP_016883469.1 | 14 | 113 | |||
GenBank mRNA | AA903471.1 | - | 1 | 258 | 113 | |
AB044943.1 | - | 16 | 113 | |||
AF308609.1 | - | 15 | 113 | |||
AK027665.1 | - | 15 | 113 | |||
AK128068.1 | - | 1 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
