Assay Details
Gene Symbol: LPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005247446.4 | XP_005247503.1 | 1 - 2 | 90 | 90 | |
XM_011512834.2 | XP_011511136.1 | 1 - 2 | 181 | 90 | ||
XM_017006380.1 | XP_016861869.1 | 1 - 2 | 182 | 90 | ||
GenBank mRNA | BX474180.1 | - | 1 - 2 | 146 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005247446.4, XM_011512834.2, XM_017006380.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
