Assay Details
Gene Symbol: UNC13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011517681.2 | XP_011515983.1 | 3 - 4 | 6626 | 113 | |
XM_011517682.2 | XP_011515984.1 | 3 - 4 | 6627 | 113 | ||
XM_011517683.2 | XP_011515985.1 | 3 - 4 | 6622 | 113 | ||
GenBank mRNA | EF139849.1 | - | 2 - 3 | 149 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
