Assay Details
Gene Symbol: LOC102725079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131941.1 | 4 - 5 | 308 | 86 | ||
GenBank mRNA | AI024749.1 | - | 1 - 2 | 86 | ||
BX094204.1 | - | 4 - 5 | 298 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
