Assay Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB044751.1 | - | 2 | 71 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB044751.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
