Assay Details
Gene Symbol: LOC105373132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011544354.2 | XP_011542656.1 | 1 | 1674 | 78 |
Gene Symbol: DUSP5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK055963.1 | - | 1 | 344 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK055963.1, XM_011544354.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
