Assay Details
Gene Symbol: MSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000251.2 | NP_000242.1 | 7 | 1309 | 77 | |
NM_001258281.1 | NP_001245210.1 | 8 | 1125 | 77 | ||
XM_005264332.3 | XP_005264389.2 | 7 | 1256 | 77 | ||
XM_011532867.1 | XP_011531169.1 | 7 | 1256 | 77 | ||
GenBank mRNA | AK222860.1 | - | 7 | 1246 | 77 | |
AK223284.1 | - | 7 | 1219 | 77 | ||
AK296831.1 | - | 6 | 784 | 77 | ||
AK297763.1 | - | 7 | 1012 | 77 | ||
AK299667.1 | - | 6 | 644 | 77 | ||
AK304496.1 | - | 8 | 1125 | 77 | ||
AK311330.1 | - | 6 | 811 | 77 | ||
BC021566.1 | - | 7 | 1198 | 4110354 | 77 | |
BX649122.1 | - | 7 | 1204 | 77 | ||
DQ648895.1 | - | 3 | 271 | 77 | ||
L47574.1 | - | 7 | 1224 | 77 | ||
L47575.1 | - | 7 | 1222 | 77 | ||
L47576.1 | - | 7 | 1223 | 77 | ||
L47577.1 | - | 7 | 1224 | 77 | ||
L47578.1 | - | 7 | 1224 | 77 | ||
L47579.1 | - | 7 | 1224 | 77 | ||
L47580.1 | - | 7 | 1224 | 77 | ||
L47581.1 | - | 7 | 1224 | 77 | ||
L47582.1 | - | 7 | 1224 | 77 | ||
L47583.1 | - | 7 | 1224 | 77 | ||
U03911.1 | - | 7 | 1187 | 77 | ||
U04045.1 | - | 7 | 1252 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
