Assay Details
Gene Symbol: CDC25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287582.1 | NP_001274511.1 | 7 | 860 | 108 | |
NM_001287583.1 | NP_001274512.1 | 7 | 977 | 108 | ||
NM_001318098.1 | NP_001305027.1 | 7 | 981 | 108 | ||
NM_001790.4 | NP_001781.2 | 7 | 864 | 108 | ||
NM_022809.3 | NP_073720.1 | 4 | 645 | 108 | ||
XM_005272145.3 | XP_005272202.1 | 7 | 977 | 108 | ||
XM_006714739.3 | XP_006714802.1 | 6 | 575 | 108 | ||
XM_011543759.2 | XP_011542061.1 | 8 | 648 | 108 | ||
XM_011543760.2 | XP_011542062.1 | 7 | 977 | 108 | ||
XM_011543761.2 | XP_011542063.1 | 4 | 758 | 108 | ||
XM_011543762.1 | XP_011542064.1 | 4 | 558 | 108 | ||
XM_011543763.1 | XP_011542065.1 | 5 | 273 | 108 | ||
XM_011543764.1 | XP_011542066.1 | 4 | 326 | 108 | ||
XM_017010101.1 | XP_016865590.1 | 7 | 855 | 108 | ||
GenBank mRNA | AF277723.1 | - | 5 | 360 | 108 | |
AF277724.1 | - | 4 | 326 | 108 | ||
AF277725.1 | - | 4 | 321 | 108 | ||
AF277726.1 | - | 3 | 231 | 108 | ||
AF312681.1 | - | 2 | 107 | 108 | ||
AJ304504.1 | - | 3 | 324 | 108 | ||
AK097710.1 | - | 7 | 977 | 108 | ||
AK301828.1 | - | 8 | 927 | 108 | ||
BC019089.2 | - | 7 | 754 | 4858949 | 108 | |
M34065.1 | - | 7 | 753 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011543764.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
