Assay Details
Gene Symbol: LINC01004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039981.2 | 1 | 947 | 65 | ||
GenBank mRNA | AK126130.1 | - | 1 | 429 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
