Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243076.2 | NP_001230005.1 | 7 | 1818 | 94 | |
NM_001949.4 | NP_001940.1 | 7 | 2402 | 94 | ||
XM_005248865.4 | XP_005248922.1 | 7 | 1896 | 94 | ||
XM_005248866.4 | XP_005248923.1 | 7 | 2625 | 94 | ||
XM_011514324.2 | XP_011512626.1 | 7 | 2754 | 94 | ||
XM_011514328.2 | XP_011512630.1 | 7 | 2658 | 94 | ||
XM_017010330.1 | XP_016865819.1 | 7 | 2681 | 94 | ||
GenBank mRNA | BC099908.1 | - | 5 | 1482 | 94 | |
BC114554.1 | - | 8 | 1823 | 94 | ||
CR749285.1 | - | 7 | 1694 | 94 | ||
D38550.1 | - | 3 | 1202 | 94 |
Gene Symbol: SLFN12L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017024006.1 | XP_016879495.1 | 5 | 94 | ||
XM_017024007.1 | XP_016879496.1 | 3 | 94 | |||
XM_017024008.1 | XP_016879497.1 | 4 | 94 | |||
XM_017024009.1 | XP_016879498.1 | 4 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017024006.1, XM_017024007.1, XM_017024008.1, XM_017024009.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
