Assay Details
Gene Symbol: C9orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287391.1 | NP_001274320.1 | 1 - 2 | 99 | 90 | |
GenBank mRNA | DB077276.1 | - | 1 - 2 | 100 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287391.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
