Assay Details
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318857.1 | NP_001305786.1 | 1 - 2 | 588 | 92 | |
NM_001318858.1 | NP_001305787.1 | 1 - 2 | 588 | 92 | ||
NR_134891.1 | 1 - 2 | 588 | 92 | |||
GenBank mRNA | FY210916.1 | - | 1 - 2 | 121 | 92 | |
FY213370.1 | - | 1 - 2 | 120 | 92 | ||
FY213777.1 | - | 1 - 2 | 121 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
