Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000310.3 | NP_000301.1 | 2 - 3 | 473 | 87 | |
XM_005271008.1 | XP_005271065.1 | 2 - 3 | 257 | 87 | ||
GenBank mRNA | AK301682.1 | - | 2 - 3 | 143 | 87 | |
AK310799.1 | - | 2 - 3 | 255 | 87 | ||
AK312287.1 | - | 2 - 3 | 248 | 87 | ||
BC008426.1 | - | 2 - 3 | 253 | 4249991 | 87 | |
BX441569.2 | - | 2 - 3 | 706 | 87 | ||
CR542053.1 | - | 2 - 3 | 241 | 87 | ||
DA504318.1 | - | 2 - 3 | 473 | 87 | ||
U44772.1 | - | 2 - 3 | 254 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
