Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016620.3 | NP_057704.2 | 2 - 3 | 294 | 91 | |
NM_032186.4 | NP_115562.3 | 2 - 3 | 324 | 91 | ||
XM_017002492.1 | XP_016857981.1 | 3 - 4 | 1314 | 91 | ||
XM_017002493.1 | XP_016857982.1 | 3 - 4 | 660 | 91 | ||
XM_017002494.1 | XP_016857983.1 | 3 - 4 | 437 | 91 | ||
GenBank mRNA | AK291520.1 | - | 2 - 3 | 94 | 91 | |
BC050656.2 | - | 2 - 3 | 243 | 91 | ||
BC110841.1 | - | 2 - 3 | 142 | 91 | ||
BI760689.1 | - | 290 | 91 | |||
DA147194.1 | - | 2 - 3 | 296 | 91 | ||
DB001551.1 | - | 2 - 3 | 433 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK291520.1, BC050656.2, BC110841.1, NM_016620.3, NM_032186.4, XM_017002492.1, XM_017002493.1, XM_017002494.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
