Assay Details
Gene Symbol: USP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080481.1 | NP_001073950.1 | 4 - 5 | 912 | 110 | |
XM_005267169.3 | XP_005267226.1 | 4 - 5 | 515 | 110 | ||
XM_005267170.4 | XP_005267227.1 | 4 - 5 | 484 | 110 | ||
XM_005267172.3 | XP_005267229.1 | 4 - 5 | 484 | 110 | ||
XM_005267175.3 | XP_005267232.1 | 4 - 5 | 483 | 110 | ||
XM_011536200.2 | XP_011534502.1 | 4 - 5 | 1742 | 110 | ||
XM_017011382.1 | XP_016866871.1 | 4 - 5 | 909 | 110 | ||
XM_017011383.1 | XP_016866872.1 | 4 - 5 | 435 | 110 | ||
XM_017011384.1 | XP_016866873.1 | 4 - 5 | 412 | 110 | ||
XM_017011385.1 | XP_016866874.1 | 4 - 5 | 483 | 110 | ||
XM_017011386.1 | XP_016866875.1 | 4 - 5 | 483 | 110 | ||
GenBank mRNA | AJ583819.2 | - | 3 - 4 | 379 | 110 | |
AK310643.1 | - | 4 - 5 | 416 | 110 | ||
AL713747.2 | - | 2 - 3 | 278 | 110 | ||
BC005991.1 | - | 4 - 5 | 430 | 110 | ||
BC150648.2 | - | 4 - 5 | 479 | 9057019 | 110 | |
BC157838.1 | - | 4 - 5 | 479 | 9057057 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AL713747.2, XM_017011382.1, XM_017011383.1, XM_017011384.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
