Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142604.1 | NP_001136076.1 | 1 - 2 | 355 | 108 | |
GenBank mRNA | AK302232.1 | - | 1 - 2 | 137 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
