Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199742.1 | NP_001186671.1 | 2 - 3 | 460 | 83 | |
GenBank mRNA | EF581092.1 | - | 2 - 3 | 423 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
