Assay Details
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005613.3 | NP_001005613.1 | 1 - 2 | 638 | 82 | |
GenBank mRNA | AF061193.1 | - | 1 - 2 | 638 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
