Assay Details
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005022.3 | NP_005013.1 | 1 - 2 | 819 | 127 | |
XM_017024761.1 | XP_016880250.1 | 1 - 2 | 821 | 127 | ||
GenBank mRNA | AK312168.1 | - | 1 - 2 | 265 | 127 | |
BC002475.2 | - | 1 - 2 | 249 | 3349355 | 127 | |
BC006768.1 | - | 1 - 2 | 242 | 2900010 | 127 | |
BC013439.1 | - | 1 - 2 | 242 | 3896948 | 127 | |
BC015164.1 | - | 1 - 2 | 257 | 3890384 | 127 | |
BC057828.1 | - | 1 - 2 | 268 | 30337970 | 127 | |
BT007001.1 | - | 1 - 2 | 127 | 127 | ||
CR407670.1 | - | 1 - 2 | 127 | 127 | ||
GU727630.1 | - | 1 - 2 | 127 | 127 | ||
J03191.1 | - | 1 - 2 | 254 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
