Assay Details
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005022.3 | NP_005013.1 | 2 - 3 | 1015 | 97 | |
GenBank mRNA | AK312168.1 | - | 2 - 3 | 97 | ||
BC002475.2 | - | 2 - 3 | 445 | 3349355 | 97 | |
BC006768.1 | - | 2 - 3 | 438 | 2900010 | 97 | |
BC013439.1 | - | 2 - 3 | 438 | 3896948 | 97 | |
BC015164.1 | - | 2 - 3 | 453 | 3890384 | 97 | |
BC057828.1 | - | 2 - 3 | 464 | 30337970 | 97 | |
BT007001.1 | - | 2 - 3 | 323 | 97 | ||
CR407670.1 | - | 2 - 3 | 323 | 97 | ||
GU727630.1 | - | 2 - 3 | 323 | 97 | ||
J03191.1 | - | 2 - 3 | 450 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
