Assay Details
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022556.3 | NP_072050.1 | 2 - 3 | 305 | 108 | |
GenBank mRNA | AB451332.1 | - | 1 - 2 | 164 | 108 | |
AB451477.1 | - | 1 - 2 | 164 | 108 | ||
AF006060.1 | - | 2 - 3 | 176 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
