Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002188.2 | NP_002179.2 | 1 - 2 | 189 | 68 | |
GenBank mRNA | BC096139.1 | - | 1 - 2 | 189 | 40002680 | 68 |
BC096141.1 | - | 1 - 2 | 189 | 40002686 | 68 | |
L06801.1 | - | 1 - 2 | 177 | 68 | ||
X69079.1 | - | 1 - 2 | 189 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000640.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
microglial cell activation
positive regulation of immunoglobulin production
movement of cell or subcellular component
inflammatory response
immune response
signal transduction
cell-cell signaling
regulation of proton transport
positive regulation of B cell proliferation
response to lipopolysaccharide
positive regulation of connective tissue growth factor production
negative regulation of NAD(P)H oxidase activity
response to nicotine
positive regulation of tyrosine phosphorylation of Stat6 protein
positive regulation of macrophage activation
positive regulation of mast cell degranulation
response to ethanol
positive regulation of smooth muscle cell proliferation
positive regulation of protein secretion
positive regulation of release of sequestered calcium ion into cytosol
cellular response to mechanical stimulus
cellular response to cytokine stimulus
negative regulation of transforming growth factor beta production
negative regulation of neuron death
negative regulation of lung ciliated cell differentiation
positive regulation of lung goblet cell differentiation
negative regulation of complement-dependent cytotoxicity
positive regulation of pancreatic stellate cell proliferation
negative regulation of endothelial cell apoptotic process
cytokine activity
interleukin-13 receptor binding
protein binding
microglial cell activation
positive regulation of immunoglobulin production
movement of cell or subcellular component
inflammatory response
immune response
signal transduction
cell-cell signaling
regulation of proton transport
positive regulation of B cell proliferation
response to lipopolysaccharide
positive regulation of connective tissue growth factor production
negative regulation of NAD(P)H oxidase activity
response to nicotine
positive regulation of tyrosine phosphorylation of Stat6 protein
positive regulation of macrophage activation
positive regulation of mast cell degranulation
response to ethanol
positive regulation of smooth muscle cell proliferation
positive regulation of protein secretion
positive regulation of release of sequestered calcium ion into cytosol
cellular response to mechanical stimulus
cellular response to cytokine stimulus
negative regulation of transforming growth factor beta production
negative regulation of neuron death
negative regulation of lung ciliated cell differentiation
positive regulation of lung goblet cell differentiation
negative regulation of complement-dependent cytotoxicity
positive regulation of pancreatic stellate cell proliferation
negative regulation of endothelial cell apoptotic process
cytokine activity
interleukin-13 receptor binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05008876.hg.1 | TC05000640.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008876.hg.1 | ENST00000304506.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05008876.hg.1 | ENST00000468334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05008876.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05008871.hg.1 | TC05000640.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008871.hg.1 | ENST00000304506.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05008871.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05008872.hg.1 | TC05000640.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008872.hg.1 | ENST00000304506.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05008872.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159295.hg.1 | TC0500008642.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159292.hg.1 | TC0500008642.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159293.hg.1 | TC0500008642.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000304506.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000459878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000462480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000468334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000487267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | ENST00000617259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008642.hg.1 | NM_002188.2 | Refseq |
